Diamond® RealSeal™ Bottles, Rectangular, High-Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
- Safe for freezer storage down to –100 °C
- Resistant to most corrosives
- Medical grade resins meet demanding requirements
- Precision molded for consistent wall thickness and smooth surfaces
- Leakproof, linerless cap for secure closing
- Laboratory bottle quality and performance with savings associated with economy bottles
- Safe, shatter-resistant alternative to glass bottles
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accommodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These rectangular bottles store and pack efficiently to maximize space utilization.
Customers Who Bought This Also Bought