Diamond® RealSeal™ Bottles, Narrow Mouth, High Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
- Safe for freezer storage down to –100 °C
- Resistant to most corrosives
- Medical grade resins meet demanding requirements
- Precision molded for consistent wall thickness and smooth surfaces
- Leakproof, linerless cap for secure closing
- Laboratory bottle quality and performance with savings associated with economy bottles
- Safe, shatter-resistant alternative to glass bottles
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accomodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These narrow mouth bottles improve pouring of liquid contents making them perfect for storing and shipping liquids.
Customers Who Bought This Also Bought