
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1.00 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS05
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995 - 0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM
Specification Test Results
Related Information
Certificates
Search by Lot NumberAYN4_Analytical_Chemistry_-Electrochemistry EMD Millipore Sustainable Products Brochure MilliporeSigma Supelco® Titration Goes Digital
Product Literature
spec_Sodium hydroxide solution-0.1M Stability data sheet Titripac_109141
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together