


Potassium hydroxide 0.1 mol/l (0.1 N) in methanol, Titripur® volumetric solution, Supelco®
CA1.11587.1000
:
- Pack type:Glass bottle
- Size:1 L
- Cat. no.:CA1.11587.1000
- Supplier No.:1.11587.1000
:
:56.11 g/mol
:3 (6.1, 8),II
:M-Clarity™ quality level = MQ100
:215-181-3
:3286
:GHS02
:GHS06
:GHS08
:Ambient
:1310-58-3
:KOH
:MFCD00003553