
Potassium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution (max. 0.00004% Ca), Supelco®
Catalog # CA1.09107.1000
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. no.:CA1.09107.1000
- Supplier No.:1.09107.1000
Specifications
About this item
:
:56.11 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:1814
:GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553