BioClean-D™ Overboots, Ansell
Supplier: Ansell Canada
Certificates
About this item
These disposable boot covers are made of BioClean-D™, a lightweight, low linting and hard-wearing fabric (PP laminated with a PE film) which provides maximum comfort and protects against fine sprays and particles.
Low-linting
- Tie-fastenings at top and ankle
- Slip-resistant polyurethane sole
- Latex-free
- Available as sterile or nonsterile version
- Elasticated top with secure ties
- A unique barcode on the outer carton (ITF-14) allows automatic data entry into Stock Control Systems
Overboots feature a slip resistant sole and easy tie-fastenings at the top and ankle. Suitable for use in Class 10, ISO 4, M2.5 environments. Sterile garments are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: EN ISO 13034 (Type PB[6]), EN 14325, EN 1149-5, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Packaging: Sterile version: One pair per sealed plastic bag, 15 sealed bags per sealed outer bag, five sealed bags of 15 pairs per sealed carton liner, one carton liner per carton (75 pairs).
Nonsterile version: 5 bags of 30 pieces per sealed carton liner; one carton liner per carton (150 pieces).
Nonsterile version: 5 bags of 30 pieces per sealed carton liner; one carton liner per carton (150 pieces).
Specifications
- Catalog No:
- CA75982-216
- CA75972-644
- 77474-000
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Seam type:
- Bound
- Bound
- Bound