Order Entry
Canada
ContactUsLinkComponent
SP Bel-Art Secador® 4.0 Vertical Profile Clear Auto-Desiccator Cabinet with Blue End Caps, Bel-Art Products, a part of SP
SP Bel-Art Secador® 4.0 Vertical Profile Clear Auto-Desiccator Cabinet with Blue End Caps, Bel-Art Products, a part of SP
Catalog # CA47743-432DP
SP Bel-Art Secador® 4.0 Vertical Profile Clear Auto-Desiccator Cabinet with Blue End Caps, Bel-Art Products, a part of SP
Catalog # CA47743-432DP
Supplier Number:  F420741116

Specifications

  • Volume:
    1.9 cu. ft.
  • Description:
    Model 4.0, Clear, with Blue End-Caps
  • Height:
    51.8 cm (20.4")
  • Cat. no.:
    CA47743-432DP
  • Electrical:
    120 V
  • No. of Shelves:
    3
  • Supplier No.:
    F420741116

Specifications

About this item

Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® auto-desiccator cabinets.

  • Durastar® co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50%, perfect for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Automatic dehumidification and desiccant regeneration cycle operate continuously every 20 minutes
  • Color-indicating silica gel inside the desiccant regeneration module allows quick visual confirmation of desiccant status
  • Replacement shelves available

Large door with easy-to-view digital hygrometer maximizes access to interior space, includes patented seal design, tab latches and security loops for a padlock or tamper-evident seals, opens right to left. Perforated shelves facilitate circulation of dry air. Compact footprint for space saving stacking up to two units high. 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.

The patented, electronic desiccant regeneration module recharges the desiccant every 20 minutes. Compared to other auto-desiccators that regenerate once every 4 to 6 hours, the Secador® auto-desiccators attains a low %RH level on start-up and consistently maintains low %rH, especially with frequent entry to the desiccator interior.