WHEATON® MicroLiter μlplate® Covers for 96-Well Microplates, DWK Life Sciences
Catalog # 97056-606
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Silicone mat with sprayed-on PTFE barrier, Round well, pre-Cut, bulk
- Environmentally Preferable:
- Cat. no.:97056-606
- Supplier No.:07-0066N-B
Specifications
About this item
MicroLiter offers a comprehensive line of 96-well microplates for chromatography.
- Piercible covers offer secure sample containment throughout the analytical sample run
- Stabilize the sample until it’s injected or maintain a measure of stability even after the sample run has finished
- Choose from two mats, an EVA mat that stabilizes the sample yet is piercible by most instruments or a silicone mat that has the resealing qualities of standard autosampler vial septa
MicroLiter offers microplates in two configurations, round well and square well. Any round well mat will fit any round well microplate. Likewise, any square well mat will fit any square well microplate.