Order Entry
Canada
ContactUsLinkComponent
Stainless Steel Bristle and Curved Wood Handle Scratch Brush, Gordon Brush
Stainless Steel Bristle and Curved Wood Handle Scratch Brush, Gordon Brush
Catalog # 22940-638
Supplier:  Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Stainless Steel Bristle and Curved Wood Handle Scratch Brush, Gordon Brush
Catalog # 22940-638
Supplier:  Gordon Brush
Supplier Number:  414SSG
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Bristle Material:
    Stainless steel
  • Brush head Ø:
    0.012"
  • Handle Material:
    Wood
  • Description:
    Scratch Brush
  • Brush Head Width:
    3/4"
  • Cat. no.:
    22940-638
  • No. of Rows:
    4×19
  • Handle Length:
    133/4"
  • Width:
    1⅛"
  • Supplier No.:
    414SSG
  • Brush Head Length:
    53/8"
  • Trim Length:
    1⅛"

Specifications

About this item

This multi-purpose brush is very effective at uniform cleaning and polishing abrasion resistant materials.

  • Long, slightly curved hardwood handle