ContactUsLinkComponent
Nylon Bristle and Zinc Plated Handle Steel Applicator Brush, Gordon Brush®
  22940-714
Product available on GSA Advantage®
Small Business Enterprise
Nylon Bristle and Zinc Plated Handle Steel Applicator Brush, Gordon Brush®
  22940-714
 :  1010NG-12
Product available on GSA Advantage®
Small Business Enterprise
  • Bristle Material:
    Nylon
  • Handle Material:
    Steel, zinc plated
  • Description:
    Applicator Brush
  • Brush Head Width:
    1/2"
  • Cat. no.:
    22940-714
  • Handle Length:
    41/2"
  • Handle Diameter:
    3/8"
  • Supplier No.:
    1010NG-12
  • Brush Head Length:
    1/2"
  • Bristle Diameter:
    0.010"
  • Trim Length:
    ³⁄₁₆" tapered, ¼" flat
Fibers are mechanically clamped into the handle for maximum fiber retention so that no adhesives can leach out and contaminate surfaces being cleaned.
  • Two sides with two end styles (tapered and flat) to fit your needs
Handle material for the 1010 series is zinc plated steel.