Tampico Bristle and Wood Handle Parts Cleaning Brush, Gordon Brush
22940-694
Product available on GSA Advantage®
Small Business Enterprise
- Bristle Material:Tampico
- Brush head Ø:11/2"
- Handle Material:Wood
- Description:Parts Cleaning Brush
- Cat. no.:22940-694
- Handle Length:73/4"
- Supplier No.:159999T
- Trim Length:3¼"
This Tampico Bristle and Plastic Handle Parts Cleaning Brush works well for removing grease, oil and solvents from metal parts.
- Metal ferrule
- Tapered plastic handle
Highly resistent to heat and chemicals.