ContactUsLinkComponent
Cotton Potash Swab Brush, Gordon Brush®
Cotton Potash Swab Brush, Gordon Brush®
  22940-654
Product available on GSA Advantage®
Small Business Enterprise
Cotton Potash Swab Brush, Gordon Brush®
  22940-654
 :  900504
Product available on GSA Advantage®
Small Business Enterprise
  • Bristle Material:
    Cotton
  • Handle Material:
    Wood
  • Description:
    Duster Brush
  • Cat. no.:
    22940-654
  • Handle Length:
    127/8"
  • Supplier No.:
    900504
This Cotton Potash Swab Brush is an extra soft swab brush made with heavy cotton yarn.
  • Great for polishing aluminum
Heavy cotton yarn is hand sewn with nylon thread into a wooden handle.