Stiff Nylon Bristle and Acetal Handle Brush, Gordon Brush
22940-582
Product available on GSA Advantage®
Small Business Enterprise
- Bristle Material:Nylon
- Handle Material:Acetal
- Description:Toothbrush-Style Brush
- Brush Head Width:5/16"
- Cat. no.:22940-582
- No. of Rows:3×11
- Handle Length:61/4"
- Width:⅜"
- Supplier No.:33NDG-12
- Brush Head Length:11/4"
- Bristle Diameter:0.012"
- Trim Length:⁷⁄₁₆"
This stiff insulative brush has 0.012" Stiff Nylon Bristle with a Delrin (Acetal) handle.
- Popular for cleaning, scrubbing, wiping or dirt removal in delicate applications
- Can be used in an anti-static environment when submerged in liquid
- Autoclavable for cleanroom, sterile, or medical applications
The Nylon fill material is hand-laced with Nylon twine into its handle for maximum fiber retention.