Brass Bristle and Plywood Handle Scratch Brush, Gordon Brush
22940-614
Product available on GSA Advantage®
Small Business Enterprise
- Bristle Material:Brass
- Brush head Ø:0.006"
- Description:Scratch Brush
- Brush Head Width:1/8"
- Cat. no.:22940-614
- No. of Rows:1×10
- Handle Length:7 3/4"
- Width:¼"
- Supplier No.:11BPG-12
- Brush Head Length:1 1/2"
- Trim Length:⁷⁄₁₆"
Brass Bristle brush has a toothbrush style plywood handle.
- Fill material is hand-tied with stainless steel wire
Remains secure in wet environments when wood handles expand and contract.