Specifications
- Description:Endure™ Aqueous Clarifier
- Size:500 ml
- Cat. no.:10809-898
- Supplier No.:ED44
Specifications
About this item
The Endure Staining System is designed to produce precise, consistent H&E staining.
- Focus clarifier for background haze and stain removal
Staining times in hematoxylin and eosin will depend on the use and the personal preference of the diagnosing physician. CDI has guidelines that will help establish a custom protocol for your specific needs. The Endure H&E System includes Hematoxylin ED1, (or ED5 “XT” for darker staining), Eosin ED2, Bluing ED3 and Clarifier (Aqueous) ED4 or Clarifier (Alcoholic) ED6. Each component is analytically optimized for staining consistency and longevity