ContactUsLinkComponent
VWR® Plus Series Built-in Undercounter Refrigerators
undefined
VWR® Plus Series Built-in Undercounter Refrigerators
  • Catalog No:
  • 76359-220
  • 76359-222
  • Cat. No.:
  • 76359-220
  • 76359-222
  • Defrost:
  • Cycle
  • Cycle
  • Shelves/drawers:
  • 4 adjustable shelves
  • 4 adjustable shelves
  • Door type:
  • 1 solid swing door
  • 1 solid swing door
  • Electrical:
  • 115 V, 0.9 A
  • 115 V, 0.9 A
  • Shipping weight:
  • 85 kg (187 lbs)
  • 85 kg (187 lbs)
  • Environmental certification:
  • Natural hydrocarbon (HC) refrigerants
  • Natural hydrocarbon (HC) refrigerants
Frequently Bought Together