Orders LinkContactUsLinkComponent
Chromatography Flow Control Adapters, ChemScience
Chromatography Flow Control Adapters, ChemScience
Supplier:   CHEM SCIENCE INC CA
Chromatography Flow Control Adapters, ChemScience
Certificates
Chromatography Flow Control Adapters, ChemScience
Chromatography Flow Control Adapters, ChemScience
4 Options Available Below
Product Details & Documents
These flow control adapters with standard taper joint has a 0 to 4 mm high vacuum valve with metering tip to adjust air pressure.
Product Family Options
Product Information
  • [ST] joint size
    Material
    Availability
    Price
  • CA10822-682
    Chromatography Flow Control Adapter
    Chromatography Flow Control Adapter
    35/20
    Glass
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $772.61
    $772.61
    Case of 10
       $772.61
    Supplier Number: CS-C01301
  • CA10822-684
    Chromatography Flow Control Adapter
    Chromatography Flow Control Adapter
    50/30
    Glass
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $948.95
    $948.95
    Case of 10
       $948.95
    Supplier Number: CS-C01302
  • CA10823-394
    Chromatography Flow Control Adapter
    Chromatography Flow Control Adapter
    24/40
    Glass
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $518.35
    $518.35
    Case of 10
       $518.35
    Supplier Number: CS-C01324
  • CA10823-396
    Chromatography Flow Control Adapter
    Chromatography Flow Control Adapter
    45/50
    Glass
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $749.29
    $749.29
    Case of 10
       $749.29
    Supplier Number: CS-C01345