ICP-MS Internal Standard 8, VeriSpec®
Catalog # RCRV010660100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Matrix:10% HNO₃/tr. HCl
- Pack type:Plastic bottle
- Description:VeriSpec® ICP-MS Internal Standard 8
- Size:100 ml
- Cat. no.:RCRV010660100N
- Supplier No.:RV010660-100N
Specifications
About this item
8-component mixture in 10% HNO₃/tr HCl
10 ppm: ⁶Li, Sc, Ge, Rh, In, Tb, Lu, Bi
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility