ICP-MS Tuning Solution 5, VeriSpec®
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
5-Component mixture in 5% HNO₃/tr HF
10 ppm: Li, Y, Ce, Tl, Co
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Specifications
Specifications
- Pack type:Plastic bottle
- Description:VeriSpec® ICP-MS tuning solution 5
- Size:100 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



