Orders LinkContactUsLinkComponent
Wire Shelving, Eagle MHC
Wire Shelving, Eagle MHC
Supplier:   Eagle Group
Wire Shelving, Eagle MHC
Supplier:  Eagle Group
Certificates
Wire Shelving, Eagle MHC
Wire Shelving, Eagle MHC
41 Options Available Below
Product Details & Documents
Patented QuadTruss® design makes these shelves up to 25% stronger and provides a retaining ledge for increased storage stability and product retention.
  • Tapered split sleeves of high temperature resistant ABS plastic create a positive lock that becomes stronger as the load increases
  • Posts are numbered in increments of 1" (25 mm) to ensure fast and level assembly
  • Levelling feet are provided to help compensate for uneven floor surfaces
  • Shelf mat utilizes a pincer-type design with the mat wire sandwiched between the two top truss wires, adding significant strength
Numerically calibrated grooved posts, tapered plastic split sleeves and shelf collars combine to make shelving assembly a simple two-step exercise: 1) snap the split sleeves onto a post over the number of your choice and 2) slide a shelf collar over the split sleeves. A positive lock between shelf and post is created without the use of any tools.
Open wire construction promotes higher visibility, allows light to pass through the shelves, permits greater air circulation which helps reduce moisture and dust build-up, and increases the effectiveness of fire-suppression systems.
Ordering information: Posts are required and sold separately. Contact your Avantor representative for more information.
Packaging: Packed in 24" (610 mm) through 48" (1219 mm) lengths are packed four to a box. 54", 60" and 72" (1372, 1524 and 1829 mm) lengths are packed two to a box.
Product Family Options
Shelves
Product Information
  • Width
    Length
    Load capacity
    Weight
    Availability
    Price
  • 58202-000
    Chrome Wire Shelf
    Chrome Wire Shelf
    14"
    24"
    800 lbs.
    2.7 kg (6 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $204.24
    $204.24
    Log in for availability
    Case of 4
    Case of 4 - $816.96
    $816.96
    Each
       $204.24
    Case of 4
       $816.96
    Supplier Number: 1424C
  • 58202-046
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    14"
    24"
    800 lbs.
    2.7 kg (6 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $714.70
    $714.70
    Log in for availability
    Case of 4
    Case of 4 - $2,858.80
    $2,858.80
    Each
       $714.70
    Case of 4
       $2,858.80
    Supplier Number: 1424S
  • 58202-922
    Chrome Wire Shelf
    Chrome Wire Shelf
    14"
    72"
    600 lbs.
    7.7 kg (17 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $318.44
    $318.44
    Each
       $318.44
    Minimum order quantity is 1
    Supplier Number: 1472C
  • 58202-052
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    14"
    72"
    600 lbs.
    7.7 kg (17 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,287.85
    $1,287.85
    Log in for availability
    Case of 2
    Case of 2 - $2,575.70
    $2,575.70
    Each
       $1,287.85
    Case of 2
       $2,575.70
    Supplier Number: 1472S
  • 58202-054
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    18"
    24"
    800 lbs.
    3.2 kg (7 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $876.52
    $876.52
    Log in for availability
    Case of 4
    Case of 4 - $3,506.08
    $3,506.08
    Each
       $876.52
    Case of 4
       $3,506.08
    Supplier Number: 1824S
  • 58202-010
    Chrome Wire Shelf
    Chrome Wire Shelf
    18"
    30"
    800 lbs.
    3.6 kg (8 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $229.77
    $229.77
    Log in for availability
    Case of 4
    Case of 4 - $919.08
    $919.08
    Each
       $229.77
    Case of 4
       $919.08
    Supplier Number: 1830C
  • 58202-926
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    18"
    30"
    800 lbs.
    3.6 kg (8 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $828.36
    $828.36
    Each
       $828.36
    Minimum order quantity is 1
    Supplier Number: 1830S
  • 58202-012
    Chrome Wire Shelf
    Chrome Wire Shelf
    18"
    36"
    800 lbs.
    4.1 kg (9 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $238.39
    $238.39
    Log in for availability
    Case of 4
    Case of 4 - $953.56
    $953.56
    Each
       $238.39
    Case of 4
       $953.56
    Supplier Number: 1836C
  • 58202-056
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    18"
    36"
    800 lbs.
    4.1 kg (9 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $899.20
    $899.20
    Log in for availability
    Case of 4
    Case of 4 - $3,596.80
    $3,596.80
    Each
       $899.20
    Case of 4
       $3,596.80
    Supplier Number: 1836S
  • 58202-058
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    18"
    42"
    800 lbs.
    5.0 kg (11 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,074.94
    $1,074.94
    Log in for availability
    Case of 4
    Case of 4 - $4,299.76
    $4,299.76
    Each
       $1,074.94
    Case of 4
       $4,299.76
    Supplier Number: 1842S
  • 58202-076
    Eaglebrite® Zinc Wire Shelf
    Eaglebrite® Zinc Wire Shelf
    18"
    42"
    800 lbs.
    5.0 kg (11 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $269.54
    $269.54
    Log in for availability
    Case of 4
    Case of 4 - $1,078.16
    $1,078.16
    Each
       $269.54
    Case of 4
       $1,078.16
    Supplier Number: 1842Z
  • 58202-014
    Chrome Wire Shelf
    Chrome Wire Shelf
    18"
    48"
    800 lbs.
    5.5 kg (12 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $283.67
    $283.67
    Log in for availability
    Case of 4
    Case of 4 - $1,134.68
    $1,134.68
    Each
       $283.67
    Case of 4
       $1,134.68
    Supplier Number: 1848C
  • 58202-016
    Chrome Wire Shelf
    Chrome Wire Shelf
    18"
    60"
    600 lbs.
    7.7 kg (17 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $334.73
    $334.73
    Log in for availability
    Case of 2
    Case of 2 - $669.46
    $669.46
    Each
       $334.73
    Case of 2
       $669.46
    Supplier Number: 1860C
  • 58202-930
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    18"
    60"
    600 lbs.
    7.7 kg (17 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $1,267.87
    $1,267.87
    Each
       $1,267.87
    Minimum order quantity is 1
    Supplier Number: 1860S
  • 58202-018
    Chrome Wire Shelf
    Chrome Wire Shelf
    18"
    72"
    600 lbs.
    9.1 kg (20 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $400.07
    $400.07
    Log in for availability
    Case of 2
    Case of 2 - $800.14
    $800.14
    Each
       $400.07
    Case of 2
       $800.14
    Supplier Number: 1872C
  • 58202-062
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    18"
    72"
    600 lbs.
    9.1 kg (20 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,622.40
    $1,622.40
    Log in for availability
    Case of 2
    Case of 2 - $3,244.80
    $3,244.80
    Each
       $1,622.40
    Case of 2
       $3,244.80
    Supplier Number: 1872S
  • 58202-020
    Chrome Wire Shelf
    Chrome Wire Shelf
    21"
    24"
    800 lbs.
    3.6 kg (8 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $263.87
    $263.87
    Log in for availability
    Case of 4
    Case of 4 - $1,055.48
    $1,055.48
    Each
       $263.87
    Case of 4
       $1,055.48
    Supplier Number: 2124C
  • 58202-932
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    21"
    24"
    800 lbs.
    3.6 kg (8 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $941.09
    $941.09
    Each
       $941.09
    Minimum order quantity is 1
    Supplier Number: 2124S
  • 58202-022
    Chrome Wire Shelf
    Chrome Wire Shelf
    24"
    24"
    800 lbs.
    4.1 kg (9 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $278.01
    $278.01
    Log in for availability
    Case of 4
    Case of 4 - $1,112.04
    $1,112.04
    Each
       $278.01
    Case of 4
       $1,112.04
    Supplier Number: 2424C
  • 58202-080
    Eaglegard® Hybrid Epoxy With Microgard®, Wire Shelf
    Eaglegard® Hybrid Epoxy With Microgard®, Wire Shelf
    24"
    24"
    800 lbs.
    4.1 kg (9 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $340.37
    $340.37
    Log in for availability
    Case of 4
    Case of 4 - $1,361.48
    $1,361.48
    Each
       $340.37
    Case of 4
       $1,361.48
    Supplier Number: 2424E
  • 58202-064
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    24"
    24"
    800 lbs.
    4.1 kg (9 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,094.84
    $1,094.84
    Log in for availability
    Case of 4
    Case of 4 - $4,379.36
    $4,379.36
    Each
       $1,094.84
    Case of 4
       $4,379.36
    Supplier Number: 2424S
  • 58202-078
    Eaglebrite® Zinc Wire Shelf
    Eaglebrite® Zinc Wire Shelf
    24"
    48"
    800 lbs.
    7.3 kg (16 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $331.96
    $331.96
    Log in for availability
    Case of 4
    Case of 4 - $1,327.84
    $1,327.84
    Each
       $331.96
    Case of 4
       $1,327.84
    Supplier Number: 2448Z
  • 58202-034
    Chrome Wire Shelf
    Chrome Wire Shelf
    24"
    72"
    600 lbs.
    11.8 kg (26 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $499.36
    $499.36
    Log in for availability
    Case of 2
    Case of 2 - $998.72
    $998.72
    Each
       $499.36
    Case of 2
       $998.72
    Supplier Number: 2472C
  • 58202-072
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    24"
    72"
    600 lbs.
    11.8 kg (26 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $2,062.15
    $2,062.15
    Log in for availability
    Case of 2
    Case of 2 - $4,124.30
    $4,124.30
    Each
       $2,062.15
    Case of 2
       $4,124.30
    Supplier Number: 2472S
  • 58202-042
    Chrome Wire Shelf
    Chrome Wire Shelf
    30"
    72"
    600 lbs.
    13.6 kg (30 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $660.87
    $660.87
    Log in for availability
    Case of 2
    Case of 2 - $1,321.74
    $1,321.74
    Each
       $660.87
    Case of 2
       $1,321.74
    Supplier Number: 3072C
  • 58202-942
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    30"
    72"
    600 lbs.
    13.6 (30 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $2,155.38
    $2,155.38
    Each
       $2,155.38
    Minimum order quantity is 1
    Supplier Number: 3072S
  • 58202-044
    Chrome Wire Shelf
    Chrome Wire Shelf
    36"
    72"
    600 lbs.
    19.5 kg (43 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $757.36
    $757.36
    Log in for availability
    Case of 2
    Case of 2 - $1,514.72
    $1,514.72
    Each
       $757.36
    Case of 2
       $1,514.72
    Supplier Number: 3672C
  • 58202-074
    Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    36"
    72"
    600 lbs.
    19.5 kg (43 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $2,703.17
    $2,703.17
    Log in for availability
    Case of 2
    Case of 2 - $5,406.34
    $5,406.34
    Each
       $2,703.17
    Case of 2
       $5,406.34
    Supplier Number: 3672S
Posts
Product Information
  • Width
    Length
    Load capacity
    Weight
    Availability
    Price
  • 58202-292
    Chrome Mobile Post
    Chrome Mobile Post
    33"
    0.9 kg (2 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $56.33
    $56.33
    Each
       $56.33
    Supplier Number: CP33-C
  • 58202-296
    Chrome Mobile Post
    Chrome Mobile Post
    63"
    1.6 kg (3.5 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $81.68
    $81.68
    Each
       $81.68
    Supplier Number: CP63-C
  • 58202-300
    Chrome Mobile Post
    Chrome Mobile Post
    74"
    1.8 kg (4 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $87.39
    $87.39
    Each
       $87.39
    Supplier Number: CP74-C
  • 58202-304
    Chrome Mobile Post
    Chrome Mobile Post
    86"
    2.3 kg (5 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $107.07
    $107.07
    Each
       $107.07
    Supplier Number: CP86-C
  • 58202-204
    Eaglegard® Hybrid Epoxy With Microgard® Post
    Eaglegard® Hybrid Epoxy With Microgard® Post
    86"
    2.3 kg (5 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $150.32
    $150.32
    Log in for availability
    Case of 4
    Case of 4 - $601.28
    $601.28
    Each
       $150.32
    Case of 4
       $601.28
    Supplier Number: P86-E
  • 58202-198
    Eaglegard® Hybrid Epoxy With Microgard®, Post
    Eaglegard® Hybrid Epoxy With Microgard®, Post
    74"
    1.8 kg (4 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $130.48
    $130.48
    Log in for availability
    Case of 4
    Case of 4 - $521.92
    $521.92
    Each
       $130.48
    Case of 4
       $521.92
    Supplier Number: P74-E
  • 58202-294
    Stainless Steel Mobile Post
    Stainless Steel Mobile Post
    54"
    1.4 kg (3 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $202.84
    $202.84
    Each
       $202.84
    Supplier Number: CP54-S
  • 58202-298
    Stainless Steel Mobile Post
    Stainless Steel Mobile Post
    63"
    1.6 kg (3.5 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $228.21
    $228.21
    Each
       $228.21
    Supplier Number: CP63-S
  • 58202-302
    Stainless Steel Mobile Post
    Stainless Steel Mobile Post
    74"
    1.8 kg (4 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $256.40
    $256.40
    Each
       $256.40
    Supplier Number: CP74-S
  • 58202-190
    Stainless Steel Post
    Stainless Steel Post
    54"
    1.4 kg (3 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.39
    $218.39
    Log in for availability
    Case of 4
    Case of 4 - $873.56
    $873.56
    Each
       $218.39
    Case of 4
       $873.56
    Supplier Number: P54-S
  • 58202-194
    Stainless Steel Post
    Stainless Steel Post
    63"
    1.6 kg (3 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $243.89
    $243.89
    Log in for availability
    Case of 4
    Case of 4 - $975.56
    $975.56
    Each
       $243.89
    Case of 4
       $975.56
    Supplier Number: P63-S
  • 58202-200
    Stainless Steel Post
    Stainless Steel Post
    74"
    1.8 kg (4 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $278.01
    $278.01
    Log in for availability
    Case of 4
    Case of 4 - $1,112.04
    $1,112.04
    Each
       $278.01
    Case of 4
       $1,112.04
    Supplier Number: P74-S
  • 58202-206
    Stainless Steel Post
    Stainless Steel Post
    86"
    2.3 kg (5 lbs.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $314.95
    $314.95
    Log in for availability
    Case of 4
    Case of 4 - $1,259.80
    $1,259.80
    Each
       $314.95
    Case of 4
       $1,259.80
    Supplier Number: P86-S