Selenium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Catalog # RCRV010414100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:10000 µg/ml
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Size:100 ml
- Cat. no.:RCRV010414100N
- Element:Selenium
- Supplier No.:RV010414-100N
Specifications
About this item
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml