Order Entry
Canada
ContactUsLinkComponent
 
Lithium Standard, for ICP, VeriSpec®, Ricca Chemical Company
  RCRV010462100N
 :  
undefined
Lithium Standard, for ICP, VeriSpec®, Ricca Chemical Company
  RCRV010462100N
 :  RV010462-100N
 :  

 

  • Concentration:
    10000 µg/ml
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010462100N
  • Element:
    Lithium
  • Supplier No.:
    RV010462-100N

 

 

  • NIST traceable
 : Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
 : 1000 ppm =1 g/L= 1000 µg/ml