Order Entry
Canada
ContactUsLinkComponent
One Bay Tote Box Carrier, Eagle MHC
One Bay Tote Box Carrier, Eagle MHC
Catalog # 58202-440
Supplier:  Eagle Group
undefined
One Bay Tote Box Carrier, Eagle MHC
Catalog # 58202-440
Supplier:  Eagle Group
Supplier Number:  TBC1-R

Specifications

  • Width:
    26"
  • Description:
    One-bay Tote Box Carrier, Non-adjustable, Resilient Casters
  • Length:
    22⅝"
  • Cat. no.:
    58202-440
  • Weight:
    55 lbs.
  • Supplier No.:
    TBC1-R

Specifications

About this item

Comes standard with three sets of tote box slides constructed of 16 gauge chrome plated steel with a channel opening of 11/8" (29 mm).

  • Accommodates a wide variety of containers

Each carrier comes standard with casters, two brake and two swivel.