About this item
Multiple use universal caps.
- Ideal for tubes 12 to 16 mm
- Made of polyethylene
- Eight colors available
Designed for easy-on, easy-off use. Cap removal is simple and quick. Evacuated glass blood collection tubes. Precision molded from low-density polyethylene, FitsAll™ fits firmly on the tube. FitsAll™ caps are very sturdy under most rigorous procedures, including centrifugation. Not suitable for autoclaving.