Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Application:qPCR dye
- Description:igScript™ SYBR Green qPCR 2X Master Mix
- Cat. no.:104013-374
- No. of Reactions:2000× 20 µl
Specifications
About this item
SYBR green qPCR 2X master mix is ideal for high-throughput real-time PCR screening and validation.
- Enhanced efficiency, specificity, and sensitivity
- Compatible with all real-time PCR instruments
- Superior gene expression results under various cycling conditions
- Robust and active for cDNA synthesis at temperatures up to 55 °C
- Useful for gene expression data validation, absolute quantification, mutation detection, pathogen detection, viral detection, GMO characterization and more
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
SYBR green qPCR 2X master mix is a ready-to-use cocktail containing all components except primers and template, for the amplification and detection of DNA in qPCR. The ig SYBR® green qPCR 2X master mix with integrated chemically-modified hot start Taq DNA polymerase, SYBR® green I fluorescent dye, ROX dye, MgCl₂, dNTPs and stabilizers. This master mix is ideal for high-throughput real-time PCR screening and validation. The amplification step features a high quality hot start Taq DNA polymerase which offers higher fidelity and better amplification.