ContactUsLinkComponent
Verification Tester for 724 and 725 Monitors, SCS, Desco Industries
Verification Tester for 724 and 725 Monitors, SCS, Desco Industries
Catalog # 76253-630
Supplier:  Menda
undefined
Verification Tester for 724 and 725 Monitors, SCS, Desco Industries
Catalog # 76253-630
Supplier:  Menda
Supplier Number:  770065
Specifications
  • Description:
    Verification Tester for 724 and 725 Monitors
  • Cat. no.:
    76253-630
Specifications
About this item
Verification tester checks the limits of the SCS 724 workstation monitor and 725 portable wrist strap monitor and comes with new ground clip and toggle switch.
  • Provides calibration verification
  • Includes ground clip contacts and toggle switch
  • NIST calibrated with certificate included
Calibration verification checks the operator test limits of the SCS 724 workstation monitor and the test limits of the 725 portable wrist strap monitor without removing them from their workstations. Ground clip contacts and toggle switch verifies the ground circuit in the 725 portable wrist strap monitor.
Certifications: Calibrated with accepted procedures and standards traceable to the National Institute of Standards and Technology.