ContactUsLinkComponent
Dual-Wire Elastic Wristbands, Desco
Dual-Wire Elastic Wristbands, Desco
Catalogue # 76239-236
Fournisseur:  Menda
undefined
Dual-Wire Elastic Wristbands, Desco
Catalogue # 76239-236
Fournisseur:  Menda
Supplier Number:  19691
Spécifications
  • Description:
    Dual-Wire Elastic Wristband
  • Cat. no.:
    76239-236
Spécifications
À propos de cet article
Adjustable elastic wristbands continuously verify the proper resistance to ground of the operator.
  • Solid stainless steel wrist band stud machined to exacting tolerances and precise profile
  • Durable, lightweight fabric in an adjustable design (with locking clasp)
  • Silver impregnated nylon knitted on inside for superior conductivity
  • Made with pure silver irreversibly bound to fibers providing both antimicrobial and anti-odor performance
  • Non-shedding, non-sloughing band material
  • Date coded providing lot traceability to ensure quality control
  • Lifetime warranty on wrist band stud
Dual wire wrist strap provides redundancy, even if one dual wire wrist strap conductor is severed the operator has a reliable path-to-ground with other wire. Use with resistive loop technology for continuous monitoring. Solid stainless steel wrist band stud machined to exacting tolerances and precise profile, ensuring a reliable path-to-ground connection to spring loaded 4 mm stainless steel constant contact coil cord snap. Adjustment design eliminates exposed conductive 'tail' of band to provide added safety and eliminate the chance of an inappropriate ground path. Medical grade 300 series stainless steel used for all metal parts to eliminate allergic skin reactions. Non-shedding, non-sloughing band material is safe for microelectronics. Greatly exceeds ESD S1.1 Bending Life Test. Tested at over 11000000 cycles vs. the 16000 requirement.