Order Entry
Canada
ContactUsLinkComponent
durAstatic® Dissipative HDPE Bottle, with Pure-Touch Pump, Menda
durAstatic® Dissipative HDPE Bottle, with Pure-Touch Pump, Menda
Catalog # 89499-028
Supplier:  Desco
undefined
durAstatic® Dissipative HDPE Bottle, with Pure-Touch Pump, Menda
Catalog # 89499-028
Supplier:  Desco
Supplier Number:  35285

Specifications

  • Capacity:
    177 ml (6 oz.)
  • Color:
    Blue
  • Description:
    durAstatic® Dissipative HDPE Bottle with Pure-Touch Pump
  • Cat. no.:
    89499-028
  • Supplier No.:
    35285

Specifications

About this item

Dissipative HDPE durAstatic™ bottles protect valuable static sensitive printed circuitry.

  • No migratory additives
  • Embossed with ESD protective symbol
  • Vessels of 6 oz. include measurements on one side to monitor fluid level
  • Dispenses approximately 0.2 cc of liquid with each pump
  • Genuine Menda 'dish-type' liquid dispenser pumps and bottle
  • One-handed operation facilitates dispensing small amount of liquid into top dish

Available in multiple colors for easy identification in the workplace, HDPE durAstatic® bottles are dissipative and are designed to protect valuable static sensitive printed circuitry. No migratory additives are used, which reduces the chance for contamination from the bottle.

The pure-touch purity protector stops fluid from re-entering bottle, eliminating the threat of contamination of bottle contents. Genuine made in the United States of America Menda pump and bottle.

Certifications: Long lasting dissipative properties: Rtt: 10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12; <2 second charge decay, tested per IEC 61340-5-1:1998.
Delivery information: Stainless steel pump includes spring-loaded lid which will stay open or stay closed, to minimize evaporation and odors.