To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Volume:10 µl
- Description:Syringe for Varian Autosamplers
- Needle Length:50 mm
- For Use With:CP8400, CP8410, CP9010, CP9050 Autosamplers
- Supplier No.:002951
Specifications
About this item
SGE 23 cone autosampler syringes meet all fit, form and function criteria of a specific autosampler model.
- High injection volume accuracy and reproducibility
- Designed with submicron tolerances
- virtually zero dead volume
The extensive range of SGE autosampler syringes meet all fit, form and function criteria of a specific autosampler model. As minimum requirements, they meet dimensional specifications, have accuracy of better than ±2%, and are designed for worry free overnight sampling.