Orders LinkContactUsLinkComponent
Adapters, Distilling, Process Reactors, Angled, Chemglass
Adapters, Distilling, Process Reactors, Angled, Chemglass
Supplier:   Chemglass
Adapters, Distilling, Process Reactors, Angled, Chemglass
Supplier:  Chemglass
Certificates
Adapters, Distilling, Process Reactors, Angled, Chemglass
Adapters, Distilling, Process Reactors, Angled, Chemglass
6 Options Available Below
Product Details & Documents
Adapter is designed for use with flasks or reaction vessel lids with a 10 or 20° angled side neck.
  • Adapter can be used with compound angled side necks
  • Five neck vessel lids and vessel lid sets are designed for use with this adapter
Adapter has a lower inner joint and two top outer joints. Lower stopcock is either a 6 mm or a 10 mm PTFE stopcock with a 1/2" or 3/4" O.D. distillate takeoff for use with a compression fitting and TFE tubing.
Product Family Options
Product Information
  • Joint size
    Stopcock size
    Availability
    Price
  • 89028-406
    Distilling Adapter, Double, 10° Angled Side Neck, 1/2" Swagelok
    Distilling Adapter, Double, 10° Angled Side Neck, 1/2" Swagelok
    24/40
    6 mm (PTFE)
    Log in for availability
    Log in for availability
    Each
    Each - 1 260,00 $
    1 260,00 $
    Each
       1 260,00 $
    Supplier Number: CG-1235-80
  • 89028-408
    Distilling Adapter, Double, 20° Angled Side Neck, 1/2" Swagelok
    Distilling Adapter, Double, 20° Angled Side Neck, 1/2" Swagelok
    24/40
    6 mm (PTFE)
    Log in for availability
    Log in for availability
    Each
    Each - 925,00 $
    925,00 $
    Each
       925,00 $
    Supplier Number: CG-1235-81
  • 89028-410
    Distilling Adapter, Double, 10° Angled Side Neck, 1/2" Swagelok
    Distilling Adapter, Double, 10° Angled Side Neck, 1/2" Swagelok
    29/42
    6 mm (PTFE)
    Log in for availability
    Log in for availability
    Each
    Each - 1 020,00 $
    1 020,00 $
    Each
       1 020,00 $
    Supplier Number: CG-1235-84
  • 89028-412
    Distilling Adapter, Double, 20° Angled Side Neck, 1/2" Swagelok
    Distilling Adapter, Double, 20° Angled Side Neck, 1/2" Swagelok
    29/42
    6 mm (PTFE)
    Log in for availability
    Log in for availability
    Each
    Each - 1 020,00 $
    1 020,00 $
    Each
       1 020,00 $
    Supplier Number: CG-1235-85
  • 89028-414
    Distilling Adapter, Double, 10° Angled Side Neck, 3/4" Swagelok
    Distilling Adapter, Double, 10° Angled Side Neck, 3/4" Swagelok
    45/50
    10 mm (PTFE)
    Log in for availability
    Log in for availability
    Each
    Each - 1 510,00 $
    1 510,00 $
    Each
       1 510,00 $
    Supplier Number: CG-1235-88
  • 89028-416
    Distilling Adapter, Double, 20° Angled Side Neck, 3/4" Swagelok
    Distilling Adapter, Double, 20° Angled Side Neck, 3/4" Swagelok
    45/50
    10 mm (PTFE)
    Log in for availability
    Log in for availability
    Each
    Each - 1 500,00 $
    1 500,00 $
    Each
       1 500,00 $
    Supplier Number: CG-1235-89