Order Entry
Canada
ContactUsLinkComponent
TMB ELISA Buffer Kit, PeproTech
TMB ELISA Buffer Kit, PeproTech
Catalog # 76304-214
Supplier:  Peprotech
CAS Number:  
TMB ELISA Buffer Kit, PeproTech
Catalog # 76304-214
Supplier:  Peprotech
Supplier Number:  900-T00
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Description:
    ELISA Buffer Kit
  • Cat. no.:
    76304-214
  • No. of Tests:
    Kit Components Sufficient to Assay 10 Plates

Specifications

About this item

PeproTech's TMB ELISA buffer kit is specifically formulated to be used in conjunction with PeproTech's Standard TMB ELISA development kits.

  • Sterile-filtered buffers
  • Ready-to-use TMB substrate
  • Use with any ELISA Kit
  • Uncoated plates

The buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions. All the reagents have been sterile filtered to minimize assay interference and maximize shelf life. This easy-to-use ELISA buffer kit can also be purchased as a stand-alone product since the plates included are not pre-coated with capture antibody. This format allows the researcher to develop and optimize an assay for use with their own capture and detection antibodies. The actual antibody concentrations and detection ranges of the ELISA will vary.

Kit contents: 20x plate coating buffer (PBS), 1x blocking buffer, 20x sample diluent, 20x wash buffer, ready-to-use stop solution, ten sterile, uncoated 96-well ELISA plates, 50 plate sealing films, buffer handling instructions.