Orders LinkContactUsLinkComponent
Papanicolaou's solution modified EA
Papanicolaou's solution modified EA
3.8 L (1 gal.)
Catalog # CA84000-132
Supplier:   Thermo Scientific
Supplier #:  75711
Papanicolaou's solution modified EA
Catalog # CA84000-132
Supplier:  Thermo Scientific
CAS Number:  
Papanicolaou's solution modified EA
Papanicolaou's solution modified EA
3.8 L (1 gal.)
CAS Number:  
Price
$465.73
Log in for availability
Each (1US Gal)
Product Details & Documents
Control the color hues and intensities of your work with Epredia™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Modified EA is used in conjunction with OG 6 or Epredia™ Modified OG for gynecological and nongynecological specimens. This formulation is optimized to reduce precipitates that are common with other traditional EA formulations. The Thermo Scientific Modified EA staining results are most similar to Thermo Scientific EA 50 and it does not contain Bismarck Brown.
Documents
Specifications
Specifications
  • Size:
    3.8 L (1 gal.)