Flasks, Micro Volumetric, Class A, Chemglass
Supplier: Chemglass
Certificates
About this item
Class 'A' micro volumetric flasks calibrated to contain.
- Flasks have hex base for stability and V shaped interior bottom for total sample retrieval
- Flask has a GPI 13-425 top thread and is supplied complete with an open top screw cap and PTFE faced silicone septa
Ordering information: See MW-102 for replacement caps and MW-104 for replacement septa.
Frequently Bought Together