Rotary Evaporator Bump Traps, Modified, Chemglass
Catalog # 80068-118
Supplier: Chemglass
Small Business Enterprise
Specifications
- ST Joint No.:24/40 top outer, 29/42 lower inner
- Capacity:250 ml
- Description:Bump trap, rotary evaporator, modified
- Cat. no.:80068-118
- Supplier No.:CG-1322-12
Specifications
About this item
Trap provides expanded volume for reactions that have a tendency to foam excessively.
- Traps have a 24/40 top outer joint and the listed size of lower inner joint
- Trap prevents entry into the condensers
The inner vapor tube is sealed to the top of the trap to prevent entry into the condenser.