Distillation Receivers, Cow Type, Chemglass
Supplier: Chemglass
Certificates
About this item
Cow type design has three receivers 45° apart.
- Condensers with lower drip tip inner joints will protrude into the spherical section
- Permitting the distillate to fall directly into the receiving flasks
Distribution adapter has a serrated connection for attachment to vacuum source. Complete item supplied with distribution adapter, three flasks of the listed size and six stainless steel springs.
Frequently Bought Together