ContactUsLinkComponent
Rotary Evaporator Bump Traps, Elliptical, Drain Holes, Chemglass
Rotary Evaporator Bump Traps, Elliptical, Drain Holes, Chemglass
Catalog # 89166-578
Supplier:  Chemglass
Small Business Enterprise
undefined
Rotary Evaporator Bump Traps, Elliptical, Drain Holes, Chemglass
Catalog # 89166-578
Supplier:  Chemglass
Supplier Number:  CG-1321-E-10
Small Business Enterprise
Specifications
  • ST Joint No.:
    24/40 Top Outer Joint
  • Description:
    Elliptical Bump Trap, Rotary Evaporator, with Drain Holes
  • Cat. no.:
    89166-578
  • Supplier No.:
    CG-1321-E-10
Specifications
About this item
Similar to 89166-576, with the addition of solvent drain holes.
  • Traps have a 24/40 top outer joint and a 24/40 lower inner joint
  • Unique elliptical shape provides large trap volume
  • This allows the use of larger recovery flasks in a rotary evaporator
  • Two 4 mm drain holes at the base of the inner tube
Rotary evaporator bump trap is used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of 'bumping'.
Unique elliptical shape provides large trap volume while decreasing the overall length when compared to standard spherical designs.