ContactUsLinkComponent
Snyder Distillation Columns, Improved, Chemglass
Snyder Distillation Columns, Improved, Chemglass
Catalog # 80068-010
Supplier:  Chemglass
Small Business Enterprise
undefined
Snyder Distillation Columns, Improved, Chemglass
Catalog # 80068-010
Supplier:  Chemglass
Supplier Number:  CG-1304-01
Small Business Enterprise
Specifications
  • Description:
    Distilling Column, Snyder, 150 mm, Improved
  • Cat. no.:
    80068-010
  • Joint Size:
    24/40
  • Supplier No.:
    CG-1304-01
Specifications
About this item
Distillation columns featuring tree section snyder column.
  • Improved design to prevent column flooding
Column has a 24/40 top outer joint and a 24/40 lower inner joint for use with Kuderna-Danish flask.