Order Entry
Canada
Orders LinkContactUsLinkComponent
Rat Recombinant SCF (from <i>E. coli</i>)

Rat Recombinant SCF (from E. coli)

50 µg
Catalog # 76303-876
Supplier:   Peprotech
Supplier #:  400-22-50UG
Rat Recombinant SCF (from <i>E. coli</i>)
Catalog # 76303-876
Supplier:  Peprotech
CAS Number:  
Rat Recombinant SCF (from E. coli)

50 µg

CAS Number:  
Price
$961.93
Log in for availability
Each (50µG)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. The human SCF gene encodes for a 273 amino acid transmembrane protein, which contains a 25 amino acid N-terminal signal sequence, a 189 amino acid extracellular domain, a 23 amino acid transmembrane domain, and a 36 amino acid cytoplasmic domain. The secreted soluble form of SCF is generated by proteolytic processing of the membrane anchored precursor. Recombinant Rat SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.

Specifications

Specifications

  • Size:
    50 µg
  • Protein/peptide name:
    SCF
  • Protein synonyms:
    Mast Cell Growth Factor (MGF)|c-Kit Ligand|Stem Cell Factor|Steel Factor
  • Purity:
    98%
  • Endotoxin level:
    Low
  • Sequence:
    MQEICRNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV THLSVSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVAC MEENAPKNVK ESLKKPETRN FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
  • Source:
    E. coli
  • Biological activity:
    Determined by the dose-dependent stimulation of the proliferation of the human TF-1 cells. The expected ED50 for this effect is 20 - 40 ng/ml.
  • Molecular weight:
    18.4 kDa
  • Shipping temperature:
    Ambient
  • Cat. No.:
    76303-876
  • Protein/peptide type:
    Recombinant
  • Gene ID:
    60427
  • Formulation:
    Lyophilized
  • Conjugation:
    Unconjugated
  • Species:
    Rat
  • Animal-free:
    Y

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.