Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IP-10 (CXCL10) (from E. coli)
Mouse Recombinant IP-10 (CXCL10) (from E. coli)
Catalog # 76303-822
Supplier:  Peprotech
CAS Number:  
Mouse Recombinant IP-10 (CXCL10) (from E. coli)
Catalog # 76303-822
Supplier:  Peprotech
Supplier Number:  250-16-50UG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Biological activity:
    Assay #1: Determined by its ability to chemoattract IL-2 activated T cells using a concentration range of 0.1-10.0 ng/ml. Assay #2: Determined by its ability to chemoattract CXCR3-transfected HEK293 cells using a concentration range of 100-500 ng/ml.
  • Gene ID:
    15945
  • Animal-free:
    Y
  • Protein synonyms:
    gamma-Interferon Inducible Protein-10|CXCL10|crg-2
  • Protein/peptide name:
    IP-10 (CXCL10)
  • Purity:
    98%
  • Molecular weight:
    8.7 kDa
  • Sequence:
    IPLARTVRCN CIHIDDGPVR MRAIGKLEII PASLSCPRVE IIATMKKNDE QRCLNPESKT IKNLMKAFSQ KRSKRAP
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-822

Specifications

About this item

IP-10 is a CXC chemokine that signals through the CXCR3 receptor. IP-10 selectively chemoattracts Th1 lymphocytes and monocytes, and inhibits cytokine-stimulated hematopoietic progenitor cell proliferation. Additionally, it is angiostatic and mitogenic for vascular smooth muscle cells. Recombinant Murine IP-10 is an 8.7 kDa protein consisting of 77 amino acids, including the four conserved cysteine residues present in CXC chemokines.