Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IL-36RA (from E. coli)
Mouse Recombinant IL-36RA (from E. coli)
Supplier:  Peprotech
Certificates
Mouse Recombinant IL-36RA (from E. coli)
Supplier:  Peprotech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Biological activity:
    Measured by its ability to inhibit secretion of IL-8 by A431 cells in the presence of human IL-36gamma.
  • Gene ID:
    54450
  • Animal-free:
    Y
  • Protein synonyms:
    IL-1 delta|Interleukin-36RA|FIL1 delta|IL-1RP3|IL-1L1|IL-1HY1|IL-1F5|IL-1ra Homolog 1
  • Protein/peptide name:
    IL-36RA
  • Purity:
    98%
  • Molecular weight:
    16.9 kDa
  • Sequence:
    VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGV QGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient

Specifications

Frequently Bought Together