About this item
Semaphorins are a large group of structurally-related, secreted, GPI-anchored, transmembrane, cell-signaling molecules. There are 8 major classifications of Semaphorins (the first seven ordered by number, 1-7, and the eighth designated V for virus), which are characterized by the existence of a conserved 500 amino acid SEMA domain at the amino terminus. Classes 3, 4, 6, and 7 are found in vertebrates only, whilst class 5 is found in both vertebrates and invertebrates. Each class is then divided into additional subgroups based on shared structural characteristics. Semaphorins primarily function as axon growth cone guidance factors during neuronal development. Semaphorin 3A acts as a chemo-repellent to axons, and an inhibitor of the growth of axons by signaling through receptors, Neuropilin-1 and Plexin-A. PeproTech's CHO cell-derived Recombinant Human Semaphorin 3A Fc is a glycosylated, disulfide-linked homodimer of 1,976 amino acid residues, which includes the SEMA domain, immunoglobulin c2-like domain, and the C-terminal basic Arg/Lys-rich domain of the mature sequence, as well as an 8-residue N-terminal His-tag and a 230-residue C-terminal Fc region linked by two glycines. Recombinant Human Semaphorin 3A Fc has a calculated molecular weight of 226.2 kDa and therefore runs above the 200kDa marker by SDS-PAGE analysis under nonreducing conditions. When run under reducing conditions, this protein migrates as three distinct bands that, due to glycosylation, run higher than expected at apparent molecular weights of approximately 120-130 kDa, 90-100 kDa, and 35-40 kDa.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Biological activity:Determined by its ability to bind recombinant rat Neuropilin‐1 Fc Chimera in a functional ELISA assay.
- Gene ID:10371
- Animal-free:Y
- Protein synonyms:SEMA3A|SEMAD
- Protein/peptide name:Semaphorin 3A Fc
- Purity:95%
- Molecular weight:226.2 kDa
- Endotoxin level:Low
- Formulation:Lyophilized
- Shipping temperature:Ambient