Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IL-34 (from HEK293 Cells)
Human Recombinant IL-34 (from HEK293 Cells)
Catalog # 76303-798
Supplier:  Peprotech
CAS Number:  
Human Recombinant IL-34 (from HEK293 Cells)
Catalog # 76303-798
Supplier:  Peprotech
Supplier Number:  200-34-50UG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    50 µg
  • Biological activity:
    Measured by its ability to induce secretion of MCP-1 from PBMCs.
  • Gene ID:
    146433
  • Animal-free:
    Y
  • Protein synonyms:
    C16orf77|Interleukin-34
  • Protein/peptide name:
    IL-34
  • Purity:
    95%
  • Molecular weight:
    52.5 kDa
  • Sequence:
    NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP KVESVLSLLN APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHHHH
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-798

Specifications

About this item

IL-34 is a homodimeric cytokine that is expressed in a range of tissues that include spleen, heart, brain, liver, kidney, lung, ovary, thymus, testis, small intestine, prostate and colon. IL-34 is a ligand for colony-stimulating factor-1 receptor (CSF1R), which also binds to CSF-1. It specifically binds CD14+ monocytes, promotes survival and proliferation of human peripheral blood monocytes, and stimulates macrophage colony formation by human bone marrow cells. IL-34, like human CSF-1, stimulates phosphorylation of MAPK1/ERK2 and MAPK3/ERK1. Recombinant Human IL-34 is a 52.5 kDa homodimeric glycoprotein consisting of 460 amino acid residues, including a C-terminal His-Tag.