Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant FGF-9 (from <i>E. coli</i>)

Human Recombinant FGF-9 (from E. coli)

50 µg
Catalog # 76303-680
Supplier:   Peprotech
Supplier #:  100-23-50UG
Human Recombinant FGF-9 (from <i>E. coli</i>)
Catalog # 76303-680
Supplier:  Peprotech
CAS Number:  
Human Recombinant FGF-9 (from E. coli)

50 µg

CAS Number:  
Price
$1,249.32
Log in for availability
Each (50µG)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

FGF-9 is a heparin-binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-9 targets glial cells, astrocytes cells and other cells that express FGFR 1c, 2c, 3b, 3c, and 4. Recombinant Human FGF-9 is a 23.2 kDa protein consisting of 206 amino acid residues.

Specifications

Specifications

  • Size:
    50 µg
  • Protein/peptide name:
    FGF-9
  • Protein synonyms:
    GAF (Glia-activating factor)|HBGF-9|Fibroblast Growth Factor-9
  • Protein Function:
    Cytokines and Growth Factors
  • Purity:
    95%
  • Endotoxin level:
    Low
  • Sequence:
    PLGEVGNYFG VQDAVPFGNV PVLPVDSPVL LSDHLGQSEA GGLPRGPAVT DLDHLKGILR RRQLYCRTGF HLEIFPNGTI QGTRKDHSRF GILEFISIAV GLVSIRGVDS GLYLGMNEKG ELYGSEKLTQ ECVFREQFEE NWYNTYSSNL YKHVDTGRRY YVALNKDGTP REGTRTKRHQ KFTHFLPRPV DPDKVPELYK DILSQS
  • Source:
    E. coli
  • Biological activity:
    Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 10 ng/ml, corresponding to a specific activity of ≥ 1 x 10^5 units/mg.
  • Molecular weight:
    23.2 kDa
  • Shipping temperature:
    Ambient
  • Cat. No.:
    76303-680
  • Protein/peptide type:
    Recombinant
  • Gene ID:
    2254
  • Formulation:
    Lyophilized
  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Animal-free:
    Y

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.