Order Entry
Canada
ContactUsLinkComponent
Human Recombinant DKK-1 (from HEK293 Cells)
Human Recombinant DKK-1 (from HEK293 Cells)
Catalog # 76303-726
Supplier:  Peprotech
CAS Number:  
Human Recombinant DKK-1 (from HEK293 Cells)
Catalog # 76303-726
Supplier:  Peprotech
Supplier Number:  120-30-50UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    50 µg
  • Biological activity:
    Determined by its ability to inhibit mWnt3a-induced TCF/LEF-luciferase activity in reporter HEK293 cells.
  • Gene ID:
    22943
  • Animal-free:
    Y
  • Protein synonyms:
    SK|Dickkopf-1|Dickkopf-related protein-1
  • Protein/peptide name:
    DKK-1
  • Purity:
    97%
  • Molecular weight:
    25.8 kDa
  • Sequence:
    TLNSVLNSNA IKNLPPPLGG AAGHPGSAVS AAPGILYPGG NKYQTIDNYQ PYPCAEDEEC GTDEYCASPT RGGDAGVQIC LACRKRRKRC MRHAMCCPGN YCKNGICVSS DQNHFRGEIE ETITESFGND HSTLDGYSRR TTLSSKMYHT KGQEGSVCLR SSDCASGLCC ARHFWSKICK PVLKEGQVCT KHRRKGSHGL EIFQRCYCGE GLSCRIQKDH HQASNSSRLH TCQRH
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-726

Specifications

About this item

DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LPR5 and LPR6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis. Mature human DKK-1 expressed in HEK293 cells is a 35-40 kDa glycoprotein containing 235 amino acid residues. The calculated molecular weight of Recombinant Human DKK-1 expressed in HEK293 cells is 25.8 kDa.