Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant PDGF-AA (from <i>E. coli</i>)

Human Recombinant PDGF-AA (from E. coli)

1 mg
Catalog # 76081-524
Marketsource
Supplier:   Peprotech
Supplier #:  AF-100-13A-1MG
Human Recombinant PDGF-AA (from <i>E. coli</i>)
Catalog # 76081-524
Supplier:  Peprotech
Marketsource
CAS Number:  
Human Recombinant PDGF-AA (from E. coli)

1 mg

CAS Number:  
Price
$11,017.77
Log in for availability
Each (1mg)
Minimum order quantity is 1
Product Details & Documents

PDGFs are disulfide-linked dimers consisting of two 12.0-13.5 kDa polypeptide chains, designated PDGF-A and PDGF-B chains. The three naturally occurring PDGFs, PDGF-AA, PDGF-BB and PDGF-AB, are potent mitogens for a variety of cell types, including smooth muscle cells, connective tissue cells, bone and cartilage cells, and some blood cells. The PDGFs are stored in platelet alpha-granules, and are released upon platelet activation. The PDGFs are involved in a number of biological processes, including hyperplasia, chemotaxis, embryonic neuron development, and respiratory tubule epithelial cell development. Two distinct signaling receptors used by PDGFs have been identified and named PDGFR-alpha and PDGFR-beta. PDGFR-alpha is a high-affinity receptor for each of the three PDGF forms. On the other hand, PDGFR-beta interacts with only PDGF-BB and PDGF-AB. Recombinant Human PDGF-AA is a 28.5 kDa disulfide-linked homodimer of two A chains (250 total amino acids).

  • Animal-free
Specifications

Specifications

  • Size:
    1 mg
  • Protein/peptide name:
    PDGF-AA
  • Protein synonyms:
    Osteosarcoma-derived Growth Factor (ODGF)|Glioma-derived growth factor (GDGF)|Platelet-Derived Growth Factor-AA
  • Purity:
    98%
  • Endotoxin level:
    Low
  • Sequence:
    SIEEAVPAVC KTRTVIYEIP RSQVDPTSAN FLIWPPCVEV KRCTGCCNTS SVKCQPSRVH HRSVKVAKVE YVRKKPKLKE VQVRLEEHLE CACATTSLNP DYREEDTGRP RESGKKRKRK RLKPT
  • Source:
    E. coli
  • Biological activity:
    The ED50 as determined by the dose-dependent stimulation of thymidine uptake by Balb/c 3T3 cells is ≤ 1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg.
  • Molecular weight:
    28.5 kDa
  • Shipping temperature:
    Ambient
  • Cat. No.:
    76081-524
  • Protein/peptide type:
    Recombinant
  • Gene ID:
    5154
  • Formulation:
    Lyophilized
  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Animal-free:
    Y

Specifications