Anti-IgG Rabbit Polyclonal Antibody GRO-alpha/MGSA (CXCL1)
Supplier: Peprotech
Certificates
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human GRO-alpha/MGSA (CXCL1). Anti-Human GRO-alpha/MGSA (CXCL1)-specific antibody was purified by affinity chromatography employing an immobilized Human GRO-alpha/MGSA (CXCL1) matrix.
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10775-870
- 10775-872
- 10775-874
- Antigen symbol:
- IgG
- IgG
- IgG
- Antigen name:
- GRO-alpha/MGSA (CXCL1)
- GRO-alpha/MGSA (CXCL1)
- GRO-alpha/MGSA (CXCL1)
- Antigen synonyms:
- Growth Regulated Protein/Melanoma Growth Stimulatory Activity, MGSAalpha, NAP-3, GRO1
- Growth Regulated Protein/Melanoma Growth Stimulatory Activity, MGSAalpha, NAP-3, GRO1
- Growth Regulated Protein/Melanoma Growth Stimulatory Activity, MGSAalpha, NAP-3, GRO1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Gene ID:
- 2919
- 2919
- 2919
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- E.coli-derived Recombinant Human GRO-alpha/MGSA (CXCL1) (PeproTech catalog# 300-11)
- E.coli-derived Recombinant Human GRO-alpha/MGSA (CXCL1) (PeproTech catalog# 300-11)
- E.coli-derived Recombinant Human GRO-alpha/MGSA (CXCL1) (PeproTech catalog# 300-11)
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 7.8 kDa
- 7.8 kDa
- 7.8 kDa
- Purification:
- Affinity purified
- Affinity purified
- Affinity purified
- Cross adsorption:
- No
- No
- No
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at -20°C prior to reconstitution.
- Store at -20°C prior to reconstitution.
- Store at -20°C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
Specifications
Frequently Bought Together