Mini Temperature/Humidity Meter
Catalog #: 470315-060
Supplier: Avantor
Specifications
- Description:Mini Temperature/Humidity Meter
- Cat. no.:470315-060
Specifications
About this item
The mini temperature and humidity meter uses a highly accurate sensor chip to measure relative humidity and temperature.
- Dual display of temperature and humidity
- °C/°F modes
- Simultaneously hold both temperature and humidity readings
- Simultaneously hold both maximum and minimum and average readings (MAX/MIN/AVG)
- Auto-power off
- Low battery indicator
- DEW-POINT and WET-BULB temperature measurement
With dew-point and wet-bulb measurements, it is perfect for many industrial applications, laboratory and other types of work environments.
Customers Who Bought This Also Bought