Squishy Circuits, Lite Kit
Squishy Circuits Lite Kit
Catalog # 470317-840
Supplier: SQUISHY CIRCUITS STORE LLC SE
Supplier #: LITE
Jump to:
Product Details & Documents
Uses conductive play dough and insulating play dough to teach electrical circuits basics in a fun, hands-on way.
The lite kit is the most basic kit and includes everything that you need to make your first Squishy Circuits light up quickly and easily.
It is ideal for classrooms, parties, or events where every participant needs their own battery pack and wants to get started building right away.
Delivery information: Kit includes 1×Battery holder, 1×Piezoelectric buzzer, 25×LEDs (5 each red, green, white, yellow and blue), 1 to 3.5 Oz. Container of Insulating Dough, and 3 to 3.5 Oz. Containers of Conductive Dough (in Red, Green, and Blue).
Specifications
- Description:Squishy Circuits Lite Kit