Adhesive Vinyl Labels 'Danger: Class II Laser In Use', HCL Labels
Catalog # 103458-968
Supplier: HCL Label
Specifications
- Description:Preprinted adhesive labels
- Dimensions:2×4"
- Material:Vinyl, adhesive
- Cat. no.:103458-968
- Legend:Danger: Class II Laser In Use
Specifications
About this item
HCL's danger class II laser in use is made of a durable adhesive vinyl.
Use this danger class II laser label to indicate when class II lasers are in use. Do not state into beam.
Packaging: Available in packs of 25.