Order Entry
Canada
ContactUsLinkComponent
Water Bath Protective Agent, polyclear MIX 30 PLUS, PolyScience
Water Bath Protective Agent, polyclear MIX 30 PLUS, PolyScience
Catalog # 76333-932
Supplier:  Polyscience
CAS Number:  
undefined
Water Bath Protective Agent, polyclear MIX 30 PLUS, PolyScience
Catalog # 76333-932
Supplier:  Polyscience
Supplier Number:  004-300063
CAS Number:  

Specifications

  • Description:
    Water Bath Protective Agent, polyclear MIX 30 PLUS
  • Size:
    1.9 l (0.5 gal.)
  • Cat. no.:
    76333-932
  • Temperature Range:
    5...90 °C (41..194 °F)

Specifications

About this item

General purpose fluid that prevents algae growth and premature rust formation. Ideal for use in heating baths, circulators, water baths and shaking water baths.

  • For routine applications above 5 °C
  • Easy to use, no dilution required